Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

673366

Sigma-Aldrich

2,2′-[(1S,2S)-1,2-Cyclohexanediylbis[(E)-(nitrilomethylidyne)]]bis[4-(tert-butyl)-6-(1-piperidinylmethyl)phenol]

97%

Synonym(s):

N,N′-Bis[(E)-5-(tert-butyl)-2-hydroxy-3-(1-piperidinylmethyl)benzylidene]-[(1S,2S)-1,2-cyclohexanediamine]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H60N4O2
CAS Number:
Molecular Weight:
628.93
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

72-81 °C

functional group

imine

SMILES string

CC(C)(C)c1cc(CN2CCCCC2)c(O)c(\C=N\[C@H]3CCCC[C@@H]3\N=C\c4cc(cc(CN5CCCCC5)c4O)C(C)(C)C)c1

InChI

1S/C40H60N4O2/c1-39(2,3)33-21-29(37(45)31(23-33)27-43-17-11-7-12-18-43)25-41-35-15-9-10-16-36(35)42-26-30-22-34(40(4,5)6)24-32(38(30)46)28-44-19-13-8-14-20-44/h21-26,35-36,45-46H,7-20,27-28H2,1-6H3/b41-25+,42-26+/t35-,36-/m0/s1

InChI key

QWZFCHMBYWXSLV-HKPCBFNZSA-N

Application

2,2′-[(1S,2S)-1,2-Cyclohexanediylbis[(E)-(nitrilomethylidyne)]]bis[4-(tert-butyl)-6-(1-piperidinylmethyl)phenol] can be used:
  • In the synthesis of chiral Ru(II) salen complexes, which are employed in the binding studies with calf thymus DNA.[1]
  • To prepare its titanium metal complexes, which are used as catalysts in the addition of organozinc compounds to α-aldiminoesters and α-ketoesters.[2][3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization, DNA binding and cleavage studies of chiral Ru (II) salen complexes
Noor-ul HK, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 74(1), 113-119 (2009)
The first catalytic asymmetric addition of dialkylzincs to α-ketoesters
DiMauro EF and Kozlowski MC
Organic Letters, 4(22), 3781-3784 (2002)
Catalytic asymmetric addition of dialkylzinc reagents to α-aldiminoesters
Basra S, et al.
Organic Letters, 8(13), 2659-2662 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service