Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

672483

Sigma-Aldrich

(S)-(−)-1-(2,4,6-Triisopropylphenyl)ethanol

≥98.5% (HPLC)

Synonym(s):

(S)-Stericol®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H28O
CAS Number:
Molecular Weight:
248.40
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

assay

≥98.5% (HPLC)

optical purity

enantiomeric ratio: ≥98:2 (HPLC)

SMILES string

CC(C)c1cc(C(C)C)c([C@H](C)O)c(c1)C(C)C

InChI

1S/C17H28O/c1-10(2)14-8-15(11(3)4)17(13(7)18)16(9-14)12(5)6/h8-13,18H,1-7H3/t13-/m0/s1

InChI key

VYKPQIIFUMKNLF-ZDUSSCGKSA-N

Legal Information

Stericol is a registered trademark of Université Joseph Fourier

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A New Asymmetric Carbon-Carbon Bond Forming Reaction: Four-Component Stereoselective Synthesis of (Z)-4,6-Dihydroxy-3-methylalk-2-enyl Methyl Sulfones This work was supported by the Swiss National Science Foundation.
Narkevitch et al.
Angewandte Chemie (International ed. in English), 39(10), 1806-1808 (2000-08-10)
Caroline Roche et al.
The Journal of organic chemistry, 70(21), 8352-8363 (2005-10-08)
Three natural pyrrolizidines, (+)-amphorogynines A and D and (+)-retronecine, have been prepared from a common lactam intermediate. This central compound, in turn, was synthesized in diastereomerically enriched form through a highly selective [2 + 2]-cycloaddition of dichloroketene with a chiral
Hamel, M., et al.
Tetrahedron Asymmetry, 16, 3406-3415 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service