Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

672238

Sigma-Aldrich

4-[Bis-(2-chloroethyl)amino]benzaldehyde

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13Cl2NO
CAS Number:
Molecular Weight:
246.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.5% (HPLC)
99%

mp

86-90 °C

functional group

aldehyde
amine
chloro

SMILES string

ClCCN(CCCl)c1ccc(C=O)cc1

InChI

1S/C11H13Cl2NO/c12-5-7-14(8-6-13)11-3-1-10(9-15)2-4-11/h1-4,9H,5-8H2

InChI key

PXUFHXLGUJLBMI-UHFFFAOYSA-N

pictograms

Skull and crossbonesCorrosion

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service