671983
(S)-2-(Boc-amino)octanedioic acid
97%
Synonym(s):
Boc-L-2-aminosuberic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C13H23NO6
CAS Number:
Molecular Weight:
289.32
MDL number:
UNSPSC Code:
12352000
assay
≥96.5% (TLC)
97%
optical purity
ee: 99% (GC)
SMILES string
CC(C)(C)OC(=O)N[C@@H](CCCCCC(O)=O)C(O)=O
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service