Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

670952

Sigma-Aldrich

1-Ethyl-2,3-dimethylimidazolium methyl carbonate

~50% in methanol: water (2:3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16N2O3
CAS Number:
Molecular Weight:
200.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

concentration

~50% in methanol: water (2:3)

refractive index

n20/D 1.416

anion traces

bromide (Br-): ≤10 mg/kg
chloride (Cl-): ≤500 mg/kg
fluoride (F-): ≤50 mg/kg
nitrate (NO3-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤500 mg/kg

SMILES string

COC([O-])=O.CCn1cc[n+](C)c1C

InChI

1S/C7H13N2.C2H4O3/c1-4-9-6-5-8(3)7(9)2;1-5-2(3)4/h5-6H,4H2,1-3H3;1H3,(H,3,4)/q+1;/p-1

InChI key

FAFIVIBGDSQQSG-UHFFFAOYSA-M

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

121.1 °F

flash_point_c

49.5 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

CBILS© offers a simple, elegant synthetic method for the production of ionic liquids, developed by the Austrian company proionic.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service