Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

670804

Sigma-Aldrich

(−)-2,3-Bis[(2R,5R)-2,5-dimethylphospholano]maleic anhydride(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

98%

Synonym(s):

2,3-Bis[(2R,5R)-2,5-dimethyl-phospholanyl]maleic anhydride(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, catASium® M(R)Rh

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36BF4O3P2Rh
CAS Number:
Molecular Weight:
624.20
MDL number:
UNSPSC Code:
12352000

assay

≥98% (31P-NMR)
98%

SMILES string

[F-].FB(F)F.C1CC=CCCC=C1.C[C@@H]2CC[C@@H](C)[PH]23[Rh+](C)[PH]4([C@H](C)CC[C@H]4C)C5=C3C(=O)OC5=O

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

catASium is a registered trademark of Umicore AG & Co. KG

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service