670723
1-Butyl-3-methylimidazolium hydrogen carbonate solution
~50% in methanol: water (2:3)
Synonym(s):
1-Butyl-3-methylimidazolium bicarbonate, 3-Butyl-1-methylimidazolium bicarbonate
About This Item
Recommended Products
concentration
~50% in methanol: water (2:3)
anion traces
bromide (Br-): ≤10 mg/kg
chloride (Cl-): ≤200 mg/kg
fluoride (F-): ≤100 mg/kg
nitrate (NO3-): ≤10 mg/kg
SMILES string
OC([O-])=O.CCCCn1cc[n+](C)c1
InChI
1S/C8H15N2.CH2O3/c1-3-4-5-10-7-6-9(2)8-10;2-1(3)4/h6-8H,3-5H2,1-2H3;(H2,2,3,4)/q+1;/p-1
InChI key
SXUPLFPGMYCSME-UHFFFAOYSA-M
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
134.6 °F
flash_point_c
57 °C
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
CBILS© offers a simple, elegant synthetic method for the production of ionic liquids, developed by the Austrian company proionic.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service