670626
1-Ethyl-3-methylimidazolium hydrogen carbonate
~50% in methanol: water (2:3)
Synonym(s):
1-Ethyl-3-methylimidazolium bicarbonate
About This Item
concentration
~50% in methanol: water (2:3)
anion traces
bromide (Br-): ≤10 mg/kg
chloride (Cl-): ≤10 mg/kg
fluoride (F-): ≤25 mg/kg
nitrate (NO3-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg
SMILES string
[O-]OC=O.CCn1cc[n+](C)c1
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
134.6 °F
flash_point_c
57 °C
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service