670529
2-[2-(Fmoc-amino)ethoxy]ethanol
98%
Synonym(s):
[2-(2-Hydroxy-ethoxy)ethyl]carbamic acid 9H-fluoren-9-ylmethyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H21NO4
CAS Number:
Molecular Weight:
327.37
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
assay
≥97.5% (HPLC)
98%
functional group
Fmoc
Storage temp.
2-8°C
SMILES string
OCCOCCNC(=O)OCC1c2ccccc2-c3ccccc13
InChI
1S/C19H21NO4/c21-10-12-23-11-9-20-19(22)24-13-18-16-7-3-1-5-14(16)15-6-2-4-8-17(15)18/h1-8,18,21H,9-13H2,(H,20,22)
Inchi Key
DUVHQSUZTXSLMW-UHFFFAOYSA-N
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service