Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

670421

Sigma-Aldrich

(1R,2R)-trans-N-Boc-1,2-cyclopentanediamine

98%

Synonym(s):

(1R)-trans-N-Boc-1,2-diaminocyclopentane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.5% (GC)
98%

form

solid

optical activity

[α]/D -14±4°, c = 1 in 1 M HCl

optical purity

enantiomeric ratio: ≥95:5%

functional group

amine

SMILES string

CC(C)(C)OC(=O)N[C@@H]1CCC[C@H]1N

InChI

1S/C10H20N2O2/c1-10(2,3)14-9(13)12-8-6-4-5-7(8)11/h7-8H,4-6,11H2,1-3H3,(H,12,13)/t7-,8-/m1/s1

InChI key

PAXDIBGWURAVIH-HTQZYQBOSA-N

Application

(1R,2R)-trans-N-Boc-1,2-cyclopentanediamine can be used as a starting material in the synthesis of novel helical oligomers of biological importance.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Caution

may darken upon storage without loss in purity

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helical foldamer containing a combination of cyclopentane-1, 2-diamine and 2, 2-dimethylmalonic acid
Yamazaki N, et al.
The Journal of Organic Chemistry, 78(19), 9991-9994 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service