Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

670052

Sigma-Aldrich

1,3-Dimethylimidazolium hydrogen carbonate solution

~50% in methanol: water (2:3)

Synonym(s):

1,3-Dimethylimidazolium bicarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N2O3
CAS Number:
Molecular Weight:
158.16
MDL number:
UNSPSC Code:
12352100

concentration

~50% in methanol: water (2:3)

refractive index

n20/D 1.418

anion traces

bromide (Br-): ≤10 mg/kg
fluoride (F-): ≤10 mg/kg
nitrate (NO3-): ≤10 mg/kg
phosphate (PO43-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

SMILES string

OC([O-])=O.Cn1cc[n+](C)c1

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

134.6 °F

flash_point_c

57 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service