Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

669733

Sigma-Aldrich

1-Butyl-3-methylimidazolium dibutyl phosphate

≥96.0% (HPLC/T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H33N2O4P
CAS Number:
Molecular Weight:
348.42
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥96.0% (HPLC/T)

impurities

≤0.5% water

refractive index

n20/D 1.472

SMILES string

CCCCn1cc[n+](C)c1.CCCCOP([O-])(=O)OCCCC

InChI

1S/C8H15N2.C8H19O4P/c1-3-4-5-10-7-6-9(2)8-10;1-3-5-7-11-13(9,10)12-8-6-4-2/h6-8H,3-5H2,1-2H3;3-8H2,1-2H3,(H,9,10)/q+1;/p-1

InChI key

NTXQJRGQUZXUMU-UHFFFAOYSA-M

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service