Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

669121

Sigma-Aldrich

1-Butyl-3-(2-pyridinylmethyl)-1H-imidazolium hexafluorophosphate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18F6N3P
CAS Number:
Molecular Weight:
361.27
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

assay

95%

reaction suitability

reagent type: catalyst

impurities

≤5% Toluene

refractive index

n/D 1.4726

density

1.303 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.CCCCn1cc[n+](Cc2ccccn2)c1

InChI

1S/C13H18N3.F6P/c1-2-3-8-15-9-10-16(12-15)11-13-6-4-5-7-14-13;1-7(2,3,4,5)6/h4-7,9-10,12H,2-3,8,11H2,1H3;/q+1;-1

InChI key

LYRTZSJBJXWSOP-UHFFFAOYSA-N

Application

Reactant for synthesis of rhodium and iridium catalysts

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service