Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

668966

Sigma-Aldrich

(R)-(+)-2,2′-Bis(di-p-tolylphosphino)-1,1′-binaphthyl

97%

Synonym(s):

(R)-Tol-BINAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H40P2
CAS Number:
Molecular Weight:
678.78
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

254-258 °C

functional group

phosphine

SMILES string

Cc1ccc(cc1)P(c2ccc(C)cc2)c3ccc4ccccc4c3-c5c(ccc6ccccc56)P(c7ccc(C)cc7)c8ccc(C)cc8

InChI

1S/C48H40P2/c1-33-13-23-39(24-14-33)49(40-25-15-34(2)16-26-40)45-31-21-37-9-5-7-11-43(37)47(45)48-44-12-8-6-10-38(44)22-32-46(48)50(41-27-17-35(3)18-28-41)42-29-19-36(4)20-30-42/h5-32H,1-4H3

InChI key

IOPQYDKQISFMJI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst involved in studies of the role of excess phosphine ligand in enantioselective conjugate addition of ethylmagnesium bromide and α,β-unsaturated ester

Reactant serving as a precursor for:
  • Catalysts used for reductive amination of ketones
  • Rh(I)-catalyst for hydrogenation of acetamidoacrylic acid derivatives
  • Chiral platinum catalysts for asymmetric Baeyer-Villiger oxidation of cyclic ketones
  • CuI-Tol-BINAP catalysts for enantioselective Michael reactions of Grignard reagents to unsaturated esters
  • BINAP Pt Dications for cation trapping

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

We present an article concerning BINAP/SEGPHOS® Ligands and Complexes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service