Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

663743

Sigma-Aldrich

trans-1-Hexen-1-ylboronic acid pinacol ester

97%

Synonym(s):

(E)-2-(1-Hexenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-1-Hexenylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23BO2
CAS Number:
Molecular Weight:
210.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.443

bp

62-65 °C/0.5-0.6 mmHg

density

0.875 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCC\C=C\B1OC(C)(C)C(C)(C)O1

InChI

1S/C12H23BO2/c1-6-7-8-9-10-13-14-11(2,3)12(4,5)15-13/h9-10H,6-8H2,1-5H3/b10-9+

InChI key

PAOJMGKYYPHNDO-MDZDMXLPSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

195.8 °F - closed cup

flash_point_c

91 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service