Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

663328

Sigma-Aldrich

Lead hydrogen phosphate

98%

Synonym(s):

Dibasic lead phosphate, Lead monohydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbHPO4
CAS Number:
Molecular Weight:
303.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
98%
form:
powder

assay

98%

form

powder

density

5.66 g/mL at 25 °C

SMILES string

[PbH2++].OP([O-])([O-])=O

InChI

1S/H3O4P.Pb/c1-5(2,3)4;/h(H3,1,2,3,4);/q;+2/p-2

InChI key

AVVSGTOJTRSKRL-UHFFFAOYSA-L

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Han Yang et al.
Shi yan sheng wu xue bao, 36(4), 301-306 (2003-10-25)
Lead phosphate deposition technique was used to investigate the changes and characteristic distribution of ATPase activity in the paranodules of tabacoo. We found that ATPase activity was closely associated with cell type and cell developmental state, thus it was different
A G Oomen et al.
Archives of environmental contamination and toxicology, 44(1), 107-115 (2002-11-16)
For children, soil ingestion via hand-to-mouth behavior can be a main route of exposure to contaminants such as lead. The ingested lead can be mobilized from the soil and form new species during the digestion process. Speciation is known to
M Watanabe et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 31(12), 1426-1429 (1983-12-01)
The distribution of the glucose-6-phosphatase (G6Pase) activities was demonstrated by in vitro whole-body autoradiography. Relatively high activities were observed in the liver, kidney, and small intestine of mice. However, these organs did not exhibit uniform activity throughout. The activity of
Carmen Enid Martínez et al.
Environmental science & technology, 38(21), 5584-5590 (2004-12-04)
Due to their relatively low solubility, lead-phosphate minerals may control Pb solution levels at a low value in natural environments. We reportthe solubility of Pb from two lead-orthophosphate mineral suspensions (beta-Pb9(PO4)6 and PbHPO4) after aging for 3 years. Lead (Pb2+)
N Zimmermann et al.
Acta histochemica, 76(1), 97-104 (1985-01-01)
Cerium-III-ions are more and more used as capturing reagent and opaque marker for the electron microscopic localization of a number of H2O2-generating enzymes as well as phosphohydrolases. Contrary to its advantages over common lead methods in the histochemical detection of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service