662879
4-(Trifluoromethoxy)benzylboronic acid pinacol ester
97%
Synonym(s):
4,4,5,5-Tetramethyl-2-[(4-trifluoromethoxy)benzyl]-1,3,2-dioxaborolane
About This Item
assay
97%
bp
100 °C/0.2 mmHg
density
1.139 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
CC1(C)OB(Cc2ccc(OC(F)(F)F)cc2)OC1(C)C
InChI
1S/C14H18BF3O3/c1-12(2)13(3,4)21-15(20-12)9-10-5-7-11(8-6-10)19-14(16,17)18/h5-8H,9H2,1-4H3
InChI key
CQXXCBRJJSMFRZ-UHFFFAOYSA-N
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
221.0 °F - closed cup
flash_point_c
105 °C - closed cup
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service