Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

662224

Sigma-Aldrich

NanoThinks ALCO11

Synonym(s):

11-Mercapto-1-undecanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H24OS
CAS Number:
Molecular Weight:
204.37
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

concentration

5 mM in ethanol

refractive index

n20/D 1.359

density

0.790 g/mL at 25 °C

SMILES string

OCCCCCCCCCCCS

Legal Information

NanoThinks is a trademark of Sigma-Aldrich Co. LLC

comparable product

Product No.
Description
Pricing

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

53.6 °F - closed cup

flash_point_c

12 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wanyi Fu et al.
Nanoscale, 9(40), 15550-15557 (2017-10-07)
Chemical modifications bring unique properties into polymeric membranes that may have enhanced filtration or separation efficiencies, antifouling, antimicrobial activity and selectivity. However, there is a lack of nanoscale characterization of the chemical additive distribution and the impacts of chemical modifiers
M Alles et al.
Biofouling, 31(5), 469-480 (2015-07-15)
Fouling release (FR) coatings are increasingly applied as an environmentally benign alternative for controlling marine biofouling. As the technology relies on removing fouling by water currents created by the motion of ships, weakening of adhesion of adherent organisms is the
František Surman et al.
Macromolecular bioscience, 15(5), 636-646 (2015-02-04)
In the current study, well-defined polymer brushes are shown as an effective surface modification to resist the adhesion of whole blood and its components. Poly[oligo(ethylene glycol)methylether methacrylate] (poly(MeOEGMA)), poly(hydroxyethyl methacrylate) (poly(HEMA)), poly[N-(2-hydroxypropyl) methacrylamide] (poly(HPMA)), and poly(carboxybetaine acrylamide) (poly(CBAA)) brushes were
Ruby May A Sullan et al.
ACS nano, 9(2), 1448-1460 (2015-02-12)
Streptococcus mutans is a Gram-positive oral bacterium that is a primary etiological agent associated with human dental caries. In the oral cavity, S. mutans adheres to immobilized salivary agglutinin (SAG) contained within the salivary pellicle on the tooth surface. Binding
Francesco Ciriello et al.
Environmental science and pollution research international, 22(16), 12469-12478 (2015-04-24)
Airborne particulate matter (PM) contains several quinones, which are able to generate reactive oxygen species impacting on cell viability. A method able to detect and quantify PM oxidative potential, based on the cytochrome c (cyt-c) reduction by means of superoxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service