66085
2-Methyl-2-butenenitrile
technical, 50-55% (GC)
Synonym(s):
2,3-Dimethylacrylonitrile
About This Item
Recommended Products
grade
technical
concentration
50-55% (GC)
impurities
~30% 2-methyl-3-butenenitrile
refractive index
n20/D 1.417
bp
115-120 °C (lit.)
density
0.810 g/mL at 20 °C (lit.)
SMILES string
C\C=C(/C)C#N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
1.4 °F - closed cup
flash_point_c
-17 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service