Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

660086

Sigma-Aldrich

1,1′-Carbonylbisbenzotriazole preparation

40 wt. % slurry in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8N6O
CAS Number:
Molecular Weight:
264.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

reaction suitability

reaction type: C-C Bond Formation

concentration

40 wt. % slurry in H2O

bp

100 °C

SMILES string

O=C(n1nnc2ccccc12)n3nnc4ccccc34

InChI

1S/C13H8N6O/c20-13(18-11-7-3-1-5-9(11)14-16-18)19-12-8-4-2-6-10(12)15-17-19/h1-8H

InChI key

ZXYBIPTYOWWVQD-UHFFFAOYSA-N

Application

New Benzotriazole Reagents

  • Coupling reagent for solid and solution phase combinatorial synthesis of ureas based on amino acids
Reagent for tert-butoxycarbonylation of acidic substrates such phenols, aromatic and aliphatic amine hydrochlorides, and aromatic carboxylic acids in the absence of a base.[1]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 62, 11599-11599 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service