Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

658804

Sigma-Aldrich

Silver, dispersion

nanoparticles, <100 nm particle size (TEM), 10 wt. % in ethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

vapor pressure

0.05 ( 20 °C)

form

dispersion
nanoparticles

autoignition temp.

748 °F

concentration

10 wt. % in ethylene glycol

refractive index

n20/D 1.333

particle size

<100 nm (TEM)

bp

198 °C (ethylene glycol)(lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

target_organs

Kidney

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

231.8 °F - closed cup

flash_point_c

111 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service