Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

658723

Sigma-Aldrich

Boc-1-tert-butoxy-1,2-dihydroisoquinoline

95%

Synonym(s):

1-tert-Butoxy-2-butoxycarbonyl-1,2-dihydroisoquinoline, BBDI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO3
CAS Number:
Molecular Weight:
303.40
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

mp

112-116 °C

SMILES string

CC(C)(C)OC1N(C=Cc2ccccc12)C(=O)OC(C)(C)C

InChI

1S/C18H25NO3/c1-17(2,3)21-15-14-10-8-7-9-13(14)11-12-19(15)16(20)22-18(4,5)6/h7-12,15H,1-6H3

InChI key

RKFZTABXZXYIOJ-UHFFFAOYSA-N

Application

A versatile tert-butoxycarbonylation protecting reagent for a variety of acidic substrates including phenols, amine hydrochlorides, carboxylic acids, N-protected amino acids, thiols, sulfonamides, and imides.[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saito, Y. et al
Tetrahedron, 62, 11599-11599 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service