Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

658502

Sigma-Aldrich

Tris(8-hydroxyquinolinato)erbium(III)

97%

Synonym(s):

ErQ3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H18ErN3O3
CAS Number:
Molecular Weight:
599.71
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

solid

mp

>300 °C

SMILES string

O([Er](Oc1cccc2cccnc12)Oc3cccc4cccnc34)c5cccc6cccnc56

InChI

1S/3C9H7NO.Er/c3*11-8-5-1-3-7-4-2-6-10-9(7)8;/h3*1-6,11H;/q;;;+3/p-3

InChI key

DMZSOZOPQZEKNW-UHFFFAOYSA-K

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryo Nagata et al.
Advanced materials (Deerfield Beach, Fla.), e1801484-e1801484 (2018-07-06)
Harvesting of both triplets and singlets yields electroluminescence quantum efficiencies of nearly 100% in organic light-emitting diodes (OLEDs), but the production efficiency of excitons that can undergo radiative decay is theoretically limited to 100% of the electron-hole pairs. Here, breaking

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service