Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

655317

Sigma-Aldrich

Dimethyl thiophosphonate

97%

Synonym(s):

Thiophosphonic acid O,O′-dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H7O2PS
CAS Number:
Molecular Weight:
126.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

bp

59 °C/16 mmHg (lit.)

density

1.185 g/mL at 25 °C (lit.)

SMILES string

CO[PH](=S)OC

InChI

1S/C2H7O2PS/c1-3-5(6)4-2/h5H,1-2H3

InChI key

BHVWDFJQEHZKIY-UHFFFAOYSA-N

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

110.8 °F

flash_point_c

43.8 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service