655295
(−)-DIP-Chloride™ solution
65-75 wt. % in α-pinene
About This Item
concentration
65-75 wt. % in α-pinene
density
0.954 g/mL at 25 °C
SMILES string
C[C@H]1[C@@H](C[C@@H]2C[C@H]1C2(C)C)B(Cl)[C@@H]3C[C@@H]4C[C@H]([C@H]3C)C4(C)C
InChI
1S/C20H34BCl/c1-11-15-7-13(19(15,3)4)9-17(11)21(22)18-10-14-8-16(12(18)2)20(14,5)6/h11-18H,7-10H2,1-6H3/t11-,12-,13+,14+,15-,16-,17-,18-/m1/s1
InChI key
PSEHHVRCDVOTID-VMAIWCPRSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
80.6 °F - closed cup
flash_point_c
27 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service