Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

653020

Sigma-Aldrich

Methyl 2-amino-4-thiazoleacetate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2S
CAS Number:
Molecular Weight:
172.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

mp

112-116 °C (lit.)
120-124 °C

SMILES string

COC(=O)Cc1csc(N)n1

InChI

1S/C6H8N2O2S/c1-10-5(9)2-4-3-11-6(7)8-4/h3H,2H2,1H3,(H2,7,8)

InChI key

XTQKFBGFHDNUFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service