Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

652822

Sigma-Aldrich

1-(4-Chlorobenzyl)piperazine

puriss., 98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15ClN2
CAS Number:
Molecular Weight:
210.70
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

puriss.

assay

98%

refractive index

n20/D 1.5578
n20/D 1.5580 (lit.)

bp

104-106 °C/0.1 mmHg (lit.)
104-106 °C/0.1 mmHg

density

1.1396 g/mL at 25 °C
1.140 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc(CN2CCNCC2)cc1

InChI

1S/C11H15ClN2/c12-11-3-1-10(2-4-11)9-14-7-5-13-6-8-14/h1-4,13H,5-9H2

InChI key

GSJXJZOWHSTWOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Hashimoto et al.
Brain research, 590(1-2), 341-344 (1992-09-11)
The neurotoxicity of 3,4-methylenedioxymethamphetamine (MDMA) in rat brain was attenuated significantly by coadministration of several benzylpiperazines (p-nitrobenzylpiperazine, p-chlorobenzylpiperazine and 1-piperonylpiperazine), which were weak inhibitors for [3H]6-nitroquipazine binding to the 5-hydroxytryptamine (5-HT) transporter in rat brain. These results suggest that these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service