65089
(R)-(−)-2-(Methoxymethyl)pyrrolidine
≥98.0% (sum of enantiomers, GC)
Synonym(s):
O-Methyl-D-prolinol
About This Item
assay
≥98.0% (sum of enantiomers, GC)
optical activity
[α]20/D −2.4±0.3°, c = 2% in benzene
refractive index
n20/D 1.446
density
0.932 g/mL at 20 °C (lit.)
SMILES string
COC[C@H]1CCCN1
InChI
1S/C6H13NO/c1-8-5-6-3-2-4-7-6/h6-7H,2-5H2,1H3/t6-/m1/s1
InChI key
CHPRFKYDQRKRRK-ZCFIWIBFSA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
113.0 °F - closed cup
flash_point_c
45 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service