Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

650889

Sigma-Aldrich

(S,S′,R,R′)-TangPhos

Synonym(s):

(1S,1S′,2R,2R′)-1,1′-Di-tert-butyl-(2,2′)-diphospholane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H32P2
CAS Number:
Molecular Weight:
286.37
UNSPSC Code:
12352005
PubChem Substance ID:

mp

60-65 °C

SMILES string

CC(C)(C)P1CCC[C@@H]1[C@H]2CCCP2C(C)(C)C

InChI

1S/C16H32P2/c1-15(2,3)17-11-7-9-13(17)14-10-8-12-18(14)16(4,5)6/h13-14H,7-12H2,1-6H3/t13-,14-,17+,18+/m1/s1

Inchi Key

SJNUZTRUIDRSJK-KNCCTNLNSA-N

Application

Chiral Quest Phosphine Ligands for Asymmetric Hydrogenation

Catalytic ligand used in:
  • Preparation of α-borono-substituted alkylarenes by asymmetric hydroboration of styrenes catalyzed by copper(I)
  • Enantioselective synthesis of spiroindane di-Me acetic acid via hydrogenation of spiroindane di-Me acetic acid catalyzed by Ru(II)
  • Asymmetric γ-addition of thiols to allenoates
  • Rhodium-catalyzed intermolecular enantioselective hydroacylation of alkynes to give alpha- and beta-substituted unsaturated ketones by kinetic resolution
  • Stereoselective preparation of β-amino nitriles via rhodium-catalyzed asymmetric hydrogenation of amino acrylonitriles
Highly efficient in rhodium-catalyzed hydrogenation of α-dehyroamino acids and α-arylenamides,[1] β-(acylamino)acrylates,[2] itaconic acids, and enol acetates.[3] Also gives outstanding enantioselectivities in asymmetric hydroformylation reactions.[4]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Axtell, A. T. et al.
Organometallics, 25, 5003-5003 (2006)
Wenjun Tang et al.
Organic letters, 4(23), 4159-4161 (2002-11-09)
The Rh-TangPhos complex is an efficient hydrogenation catalyst for making chiral beta-amino acid derivatives. With the Rh-TangPhos system, high enantioselectivities (up to 99.6%) and turnover numbers have been obtained in the hydrogenation of E/Z isomeric mixtures of both beta-alkyl and
Wenjun Tang et al.
Organic letters, 5(2), 205-207 (2003-01-17)
[reaction: see text] The Rh-TangPhos catalyst has been used for asymmetric hydrogenation of itaconic acid and enol acetate derivatives. A variety of chiral 2-substituted succinic acids and chiral acetates have been obtained in excellent ee values (up to 99% ee).
A chiral 1,2-bisphospholane ligand with a novel structural motif: applications in highly enantioselective Rh-catalyzed hydrogenations.
Wenjun Tang et al.
Angewandte Chemie (International ed. in English), 41(9), 1612-1614 (2002-05-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service