Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

650234

Sigma-Aldrich

1-(4-Bromobenzyl)piperazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15BrN2
CAS Number:
Molecular Weight:
255.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

drug control

Home Office Schedule 1

bp

112-116 °C/0.3 mmHg (lit.)

mp

56-62 °C (lit.)

functional group

bromo

SMILES string

Brc1ccc(CN2CCNCC2)cc1

InChI

1S/C11H15BrN2/c12-11-3-1-10(2-4-11)9-14-7-5-13-6-8-14/h1-4,13H,5-9H2

InChI key

MAHWBNAOEVAPJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service