Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

649090

Sigma-Aldrich

Methyl 2-hexynoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C≡CCO2CH3
CAS Number:
Molecular Weight:
126.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.441 (lit.)

bp

64-66 °C/10 mmHg (lit.)

density

0.943 g/mL at 25 °C (lit.)

functional group

ester

storage temp.

2-8°C

SMILES string

CCCC#CC(=O)OC

InChI

1S/C7H10O2/c1-3-4-5-6-7(8)9-2/h3-4H2,1-2H3

InChI key

DHZPZBFOJDXEHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

Keep cold!

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

149.0 - 150.0 °F

flash_point_c

65 - 65.56 °C

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service