All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C28H22
CAS Number:
Molecular Weight:
358.47
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
mp
279.4-280.9 °C (lit.)
λmax
376 nm
SMILES string
Cc1ccc(cc1)-c2c3ccccc3c(-c4ccc(C)cc4)c5ccccc25
General description
Photoluminescent small molecule.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Dam. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service