Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

648914

Sigma-Aldrich

3-(1,2,3,4-Tetrahydroquinolin-2-yl)-2-(trifluoromethyl) propionic acid

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14F3NO2
CAS Number:
Molecular Weight:
273.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

90%

refractive index

n20/D 1.5093

density

1.2341 g/mL at 25 °C

SMILES string

OC(=O)C(CC1CCc2ccccc2N1)C(F)(F)F

InChI

1S/C13H14F3NO2/c14-13(15,16)10(12(18)19)7-9-6-5-8-3-1-2-4-11(8)17-9/h1-4,9-10,17H,5-7H2,(H,18,19)

InChI key

AXIQSYQEUQWCDQ-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

133.0 °F - closed cup

flash_point_c

56.1 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service