648426
(−)-DIP-Chloride™ solution
50-65 wt. % in hexanes
About This Item
Recommended Products
concentration
50-65 wt. % in hexanes
density
0.929 g/mL at 25 °C
SMILES string
C[C@H]1[C@@H](C[C@@H]2C[C@H]1C2(C)C)B(Cl)[C@@H]3C[C@@H]4C[C@H]([C@H]3C)C4(C)C
InChI
1S/C20H34BCl/c1-11-15-7-13(19(15,3)4)9-17(11)21(22)18-10-14-8-16(12(18)2)20(14,5)6/h11-18H,7-10H2,1-6H3/t11-,12-,13+,14+,15-,16-,17-,18-/m1/s1
InChI key
PSEHHVRCDVOTID-VMAIWCPRSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
signalword
Danger
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
<-22.0 °F - closed cup
flash_point_c
< -30 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service