Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

646636

Sigma-Aldrich

(S)-(+)-COP-Cl Catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C78H66Cl2Co2N2O2Pd2
CAS Number:
Molecular Weight:
1464.98
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

optical activity

[α]20/D +1201°, c = 0.1 in methylene chloride

mp

204-205 °C (lit.)

SMILES string

[Co].[Co].[Cl+]1[Pd-][Cl+][Pd-]1.CC(C)[C@@H]2COC(=N2)[C]3[CH][CH][CH][CH]3.CC(C)[C@@H]4COC(=N4)[C]5[CH][CH][CH][CH]5.c6ccc(cc6)[C]7[C]([C]([C]7c8ccccc8)c9ccccc9)c%10ccccc%10.c%11ccc(cc%11)[C]%12[C]([C]([C]%12c%13ccccc%13)c%14ccccc%14)c%15ccccc%15

InChI

1S/2C28H20.2C11H14NO.2Cl.2Co.2Pd/c2*1-5-13-21(14-6-1)25-26(22-15-7-2-8-16-22)28(24-19-11-4-12-20-24)27(25)23-17-9-3-10-18-23;2*1-8(2)10-7-13-11(12-10)9-5-3-4-6-9;;;;;;/h2*1-20H;2*3-6,8,10H,7H2,1-2H3;;;;;;/q;;;;2*+1;;;2*-1/t;;2*10-;;;;;;/m..00....../s1

InChI key

OSHCXJIQKOINDY-TWWPWAOZSA-N

Application

Catalyst employed in the rearrangement of allylic trichloracetimidates to allylic amines of high enantiopurity.[1]
Catalyst for:
  • Enantioselective allylation of phenols with (E)-allylic alcohols to branched chiral allylic aryl ethers
  • Asymmetric aza-Claisen rearrangement and ring-closing metathesis

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolyn E Anderson et al.
Journal of the American Chemical Society, 125(41), 12412-12413 (2003-10-09)
COP-Cl catalyzes the rearrangement of (E)-allylic trichloroacetimidates to provide transposed allylic trichloroacetamides of high enantiopurity, a transformation that underlies the first truly practical method for transforming prochiral allylic alcohols to enantioenriched allylic amines and congeners. The high functional group compatibility

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service