Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

644498

Sigma-Aldrich

Benzene

purification grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6
CAS Number:
Molecular Weight:
78.11
Beilstein/REAXYS Number:
969212
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
grade:
purification grade
assay:
≥99.75%
bp:
80 °C (lit.)
vapor pressure:
166 mmHg ( 37.7 °C)
74.6 mmHg ( 20 °C)
Pricing and availability is not currently available. Contact Technical Service

grade

purification grade

vapor density

2.77 (vs air)

vapor pressure

166 mmHg ( 37.7 °C)
74.6 mmHg ( 20 °C)

assay

≥99.75%

autoignition temp.

1043 °F

expl. lim.

8 %

evapn. residue

≤0.0005%

refractive index

n20/D 1.501 (lit.)

bp

80 °C (lit.)

mp

5.5 °C (lit.)

density

0.874 g/mL at 25 °C (lit.)

SMILES string

c1ccccc1

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H

InChI key

UHOVQNZJYSORNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

target_organs

Blood

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

12.2 °F

flash_point_c

-11 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sachiko Kitamoto et al.
Mutation research. Genetic toxicology and environmental mutagenesis, 786-788, 137-143 (2015-07-28)
As a part of the Japanese Center for the Validation of Alternative Methods (JaCVAM)-initiative international validation study of the in vivo alkaline comet assay (comet assay), we examined DNA damage in the liver, stomach, and bone marrow of rats dosed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service