644072
3,5-Dimethylpyrazole-1-carboxamidine hydrochloride, polymer-supported
100-200 mesh, extent of labeling: 1.0-2.0 mmol/g loading, 2 % cross-linked
All Photos(1)
About This Item
crosslinking
2 % cross-linked
extent of labeling
1.0-2.0 mmol/g loading
particle size
100-200 mesh
InChI
1S/C6H10N4.Cl/c1-4-3-5(2)10(9-4)6(7)8;/h3H,1-2H3,(H3,7,8);
InChI key
VEBVDVIHRVPBQW-UHFFFAOYSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)