Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

643106

Supelco

Water solution

contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
MDL number:
UNSPSC Code:
12190000

contains

0.1 % (v/v) trifluoroacetic acid
5 % (v/v) acetonitrile

density

0.991 g/mL at 25 °C

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Product filtered through a 0.2 μm filter

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service