Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

641782

Sigma-Aldrich

Pentaphene-78

Synonym(s):

16,17-Bis(octyloxy)-anthra[9,1,2-cde]benzo[rst]pentaphene-5,1-dione, 5-10-Dioxo-17-(stearoyloxy)-5,5a,6,10-tetrahydroanthra[9,1,2-cde]benzo[rst]pentaphen-16-yl stearate, Violanthrone-78

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H84O6
CAS Number:
Molecular Weight:
1021.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

mp

287.2 °C (dec.) (lit.)

λmax

≤577 nm in toluene

SMILES string

CCCCCCCCCCCCCCCCCC(=O)Oc1cc2-c3ccccc3C(=O)c4ccc5c6ccc7C(=O)c8ccccc8-c9cc(OC(=O)CCCCCCCCCCCCCCCCC)c(c1c5c24)c6c79

InChI

1S/C70H84O6/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-41-61(71)75-59-47-57-49-37-33-35-39-53(49)69(73)55-45-43-51-52-44-46-56-64-58(50-38-34-36-40-54(50)70(56)74)48-60(68(66(52)64)67(59)65(51)63(55)57)76-62(72)42-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33-40,43-48H,3-32,41-42H2,1-2H3

InChI key

CMDCDHZHIBRIKE-UHFFFAOYSA-N

Application

An n-channel organic semiconductor.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Newman, C.R., et al.
Chemistry of Materials, 16, 4436-4436 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service