Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

640719

Sigma-Aldrich

(1E)-1-Methoxy-2-methyl-1-penten-3-one

Synonym(s):

(E)-1-Methoxy-2-methylpent-1-en-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH=C(CH3)COCH2CH3
CAS Number:
Molecular Weight:
128.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.4770 (lit.)

bp

62 °C/6 mmHg (lit.)

density

0.961 g/mL at 25 °C (lit.)

functional group

ether
ketone

storage temp.

2-8°C

SMILES string

CCC(=O)\C(C)=C\OC

InChI

1S/C7H12O2/c1-4-7(8)6(2)5-9-3/h5H,4H2,1-3H3/b6-5+

InChI key

AIFFWMXMBCHYFL-AATRIKPKSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

177.8 °F - closed cup

flash_point_c

81 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service