Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

638978

Sigma-Aldrich

2-Ethoxybenzoyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9ClO2
CAS Number:
Molecular Weight:
184.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

bp

114-118 °C/5 mmHg (lit.)

density

1.190 g/mL at 25 °C (lit.)

SMILES string

CCOc1ccccc1C(Cl)=O

InChI

1S/C9H9ClO2/c1-2-12-8-6-4-3-5-7(8)9(10)11/h3-6H,2H2,1H3

InChI key

MDKAAWDKKBFSTK-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service