Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

637157

Sigma-Aldrich

Indium(III) hydroxide

nanopowder, <100 nm particle size, 99.99% trace metals basis

Synonym(s):

Indium trihydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In(OH)3
CAS Number:
Molecular Weight:
165.84
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

vapor pressure

<0.01 mmHg ( 25 °C)

assay

99.99% trace metals basis

form

nanopowder

reaction suitability

reagent type: catalyst
core: indium

surface area

57 m2/g

particle size

<100 nm

mp

150 °C (lit.)

density

4.45 g/mL at 20 °C (lit.)

bulk density

0.49‑0.53 g/mL

SMILES string

O[In](O)O

InChI

1S/In.3H2O/h;3*1H2/q+3;;;/p-3

InChI key

IGUXCTSQIGAGSV-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Chronic 2 - STOT RE 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service