Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

636762

Sigma-Aldrich

1,4-Bis(3,7-dimethyloctyl)-2,5-diiodobenzene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44I2
CAS Number:
Molecular Weight:
610.44
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

mp

54-55 °C (lit.)

functional group

iodo

SMILES string

CC(C)CCCC(C)CCc1cc(I)c(CCC(C)CCCC(C)C)cc1I

InChI

1S/C26H44I2/c1-19(2)9-7-11-21(5)13-15-23-17-26(28)24(18-25(23)27)16-14-22(6)12-8-10-20(3)4/h17-22H,7-16H2,1-6H3

InChI key

LJNUMLPEKWLXIO-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service