Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

635050

Sigma-Aldrich

LSAT

single crystal substrate, <100>, ≥99.9% trace metals basis

Synonym(s):

Lanthanum aluminum oxide-strontium aluminum tantalum oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(LaAlO3)3(Sr2AlTaO6)7
Molecular Weight:
3995.82
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

description

single side polished

assay

≥99.9% trace metals basis

dielectric constant

~22

size

10 mm × 10 mm × 0.5 mm

mp

1840 °C (lit.)

density

6.74 g/mL at 25 °C (lit.)

semiconductor properties

<100>

SMILES string

O=[Ta]12O[Sr]O[Al](O[Sr]O[Al]3O[Sr]O[Ta]45(O[Sr]O[Ta]67(O[Sr]O[Al](O[Sr]O[Al]8O[Sr]O[Ta]9%10(O[Sr]O[Ta]%11%12(O[Sr]O[Al](O[Sr]O[Al]%13O[Sr]O[Ta]%14%15(O[Sr]O[Ta]%16(O[Sr]O[Al](O[Sr]O1)O%16)(O[Al]%17O[La](O[Al](O[La](O[Al](O[La](O%17)O%14)O%11)O9)O6)O4)O%15)O%13)O%12)O%10)O8)O7)O5)O3)O2

InChI

1S/10Al.3La.51O.14Sr.7Ta

InChI key

YSEGFJHLRWDERW-UHFFFAOYSA-N

Physical properties

Thermal expansion = 10 × 10 -6/°C

Physical form

cubic (a = 3.868 Å)

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco J Román et al.
NeuroImage, 199, 172-183 (2019-06-04)
Although cognitive neuroscience has made remarkable progress in understanding the neural foundations of goal-directed behavior and decision making, neuroscience research on decision making competence, the capacity to resist biases in human judgment and decision making, remain to be established. Here
Jong-Cheol Rah et al.
Frontiers in neural circuits, 7, 177-177 (2013-11-26)
The subcellular locations of synapses on pyramidal neurons strongly influences dendritic integration and synaptic plasticity. Despite this, there is little quantitative data on spatial distributions of specific types of synaptic input. Here we use array tomography (AT), a high-resolution optical
Takeshi Yajima et al.
Nature chemistry, 7(12), 1017-1023 (2015-11-21)
Oxynitrides have been explored extensively in the past decade because of their interesting properties, such as visible-light absorption, photocatalytic activity and high dielectric permittivity. Their synthesis typically requires high-temperature NH3 treatment (800-1,300 °C) of precursors, such as oxides, but the highly
Drew M Morris et al.
Applied ergonomics, 63, 9-16 (2017-05-16)
Physiological tracers of circadian rhythms and a performance awareness index were examined as predictors of cognitive performance during two sleep deprivation conditions common to occupational shiftwork. Study 1: Thirty-three sleep-deprived participants completed a simulated nightshift. Study 2: Thirty-two partially sleep-deprived
Allyson P Mackey et al.
Frontiers in neuroanatomy, 6, 32-32 (2012-09-01)
Diffusion tensor imaging (DTI) techniques have made it possible to investigate white matter plasticity in humans. Changes in DTI measures, principally increases in fractional anisotropy (FA), have been observed following training programs as diverse as juggling, meditation, and working memory.

Articles

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service