Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

632139

Sigma-Aldrich

2-Methoxy-3-pyridinecarboxaldehyde

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

refractive index

n20/D 1.5500 (lit.)

bp

200-201 °C (lit.)

density

1.161 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1cccnc1OC

InChI

1S/C7H7NO2/c1-10-7-6(5-9)3-2-4-8-7/h2-5H,1H3

InChI key

PIFFMIDNNWOQLK-UHFFFAOYSA-N

Application

2-Methoxy-3-pyridinecarboxaldehyde may be used to synthesize the following:
  • ethyl 5-amino-4-(2-methoxy-3-pyridyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrano[2,3-b]quinoline-3-carboxylate, a potent AChE inhibitor[1]
  • N-hydroxy-2-pyridinone-based arylsulfone containing a Zn-binding group (ZBG) as MMP-2/-9 inhibitor[2]

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

204.8 °F - closed cup

flash_point_c

96 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis and biological evaluation of new 4H-pyrano [2, 3-b] quinoline derivatives that block acetylcholinesterase and cell calcium signals, and cause neuroprotection against calcium overload and free radicals"
Contelles-Marco J, et al.
European Journal of Organic Chemistry, 41(12), 1464-1469 (2006)
"Syntheses and in vitro evaluation of arylsulfone-based MMP inhibitors with heterocycle-derived zinc-binding groups (ZBGs)"
Zhang M-Y, et al.
Bioorganic & Medicinal Chemistry, 18(01), 405-408 (2008)
Cheuk-Fai Chow et al.
Chemistry, an Asian journal, 3(8-9), 1324-1335 (2008-07-30)
Nine neutral dynamic metallosupramolecular polymers (metallodynamers) based on acyl hydrazone derived metal coordination centers (Co(2+), Ni(2+), Zn(2+), and Cd(2+)) were generated through self-assembly polymerization. These are linear coordination polymers with specific optical and mechanical properties. Monomer selection was found to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service