Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

630632

Sigma-Aldrich

2-Phenyl-2′,4′,6′-trihydroxyacetophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O4
CAS Number:
Molecular Weight:
244.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

161-165 °C (lit.)

SMILES string

Oc1cc(O)c(c(O)c1)C(=O)Cc2ccccc2

InChI

1S/C14H12O4/c15-10-7-12(17)14(13(18)8-10)11(16)6-9-4-2-1-3-5-9/h1-5,7-8,15,17-18H,6H2

InChI key

SLHBRIIHMDJIBT-UHFFFAOYSA-N

Application

2-Phenyl-2′,4′,6′-trihydroxyacetophenone can undergo reaction with citral to give the corresponding polycycle containing a citran and cyclol nucleus, which may possess biological and pharmaceutical properties.[1]

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Efficient synthesis of polycycles by electrocyclizations of substituted trihydroxybenzenes: Synthesis of rubranine and deoxybruceol"
Lee RY and Kim HJ
Synlett, 2007(14), 2232-2236 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service