615129
Ethanol-OD
95% in D2O, 99 atom % D
Synonym(s):
Ethan(ol-d), Ethyl alcohol-OD
About This Item
isotopic purity
99 atom % D
expl. lim.
3.5-15 % (lit.)
concentration
95% in D2O
refractive index
n20/D 1.359 (lit.)
bp
78 °C (lit.)
mp
-114 °C (lit.)
density
0.806 g/mL at 25 °C
mass shift
M+1
SMILES string
[2H]OCC
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3/i3D
InChI key
LFQSCWFLJHTTHZ-WFVSFCRTSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Packaging
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
57.2 °F
flash_point_c
14 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service