Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

610275

Supelco

DL-Methamphetamine solution

1 mg/mL in methanol, drug standard

Synonym(s):

d-Deoxyephedrine solution, DL-1-Phenyl-2(methylamino)propane solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N
Molecular Weight:
149.23
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

drug standard

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

availability

not available in Japan

concentration

1 mg/mL in methanol

storage temp.

−20°C

SMILES string

CNC(C)Cc1ccccc1

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service