Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

597759

Sigma-Aldrich

Chloromethyl(dimethyl)methoxysilane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2Si(CH3)2OCH3
CAS Number:
Molecular Weight:
138.67
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

97%

form

liquid

density

0.980 g/mL at 25 °C (lit.)

SMILES string

CO[Si](C)(C)CCl

InChI

1S/C4H11ClOSi/c1-6-7(2,3)4-5/h4H2,1-3H3

InChI key

ZCSLOBFDVTWIBL-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

80.0 °F - closed cup

flash_point_c

26.67 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service