Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

591564

Sigma-Aldrich

Methyl (3R)-(−)-3-(1-methylindol-3-yl)butanoate

98%, optical purity ee: 97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO2
CAS Number:
Molecular Weight:
231.29
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

optical activity

[α]20/D −7°, c = 1% in chloroform

optical purity

ee: 97% (HPLC)

SMILES string

COC(=O)C[C@@H](C)c1cn(C)c2ccccc12

InChI

1S/C14H17NO2/c1-10(8-14(16)17-3)12-9-15(2)13-7-5-4-6-11(12)13/h4-7,9-10H,8H2,1-3H3/t10-/m1/s1

InChI key

QIOUBQLAFUPETC-SNVBAGLBSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service